Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 5-hydroxypyridine-2-carboxylate | 30766-12-2 | 153.14 | 100 G
SDP

Catalog No. 5000448589
Encompass_Preferred

Methyl 5-hydroxypyridine-2-carboxylate is an analytical standard and phenolic acid derived from *Mahonia fortune*. This compound exhibits NO inhibitory effects *in vitro* and is intended for research and analytical applications.

  • Inhibits NO in LPS-stimulated RAW264.7 and BV2 cells
  • Shows no cytotoxic effect at 6.25-200 μM concentrations
  • Appears as an off-white to pink solid
  • Soluble in DMSO (50 mg/mL)
  • Recommended storage for powder: -20°C for 3 years; 4°C for 2 years
  • Recommended storage in solvent: -80°C for 2 years; -20°C for 1 year

Catalog No. 50-004-48589 Supplier Medchemexpress LLC Supplier No. HYW005963100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 5-hydroxypyridine-2-carboxylate is an analytical standard and phenolic acid derived from *Mahonia fortune*. This compound exhibits NO inhibitory effects *in vitro* and is intended for research and analytical applications.

  • Inhibits NO in LPS-stimulated RAW264.7 and BV2 cells
  • Shows no cytotoxic effect at 6.25-200 μM concentrations
  • Appears as an off-white to pink solid
  • Soluble in DMSO (50 mg/mL)
  • Recommended storage for powder: -20°C for 3 years; 4°C for 2 years
  • Recommended storage in solvent: -80°C for 2 years; -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.