Learn More
Medchemexpress LLC Methyl 5-hydroxypyridine-2-carboxylate | 30766-12-2 | 153.14 | 25 G
SDP
Catalog No. 5000448203
Encompass_Preferred
Methyl 5-hydroxypyridine-2-carboxylate is a phenolic acid found in the stems of Mahonia fortune. It exhibits NO inhibitory effects in vitro, inhibiting NO in LPS-stimulated RAW264.7 and BV2 cells, with IC50s of 115.67 and 118.80 μM, respectively. It shows no cytotoxic effect at concentrations from 6.25-200 μM.
  • Phenolic acid
  • Exhibits NO inhibitory effects in vitro
  • Not cytotoxic at specified concentrations
Catalog No. 50-004-48203 Supplier Medchemexpress LLC Supplier No. 25GHYW005963
Methyl 5-hydroxypyridine-2-carboxylate is a phenolic acid found in the stems of Mahonia fortune. It exhibits NO inhibitory effects in vitro, inhibiting NO in LPS-stimulated RAW264.7 and BV2 cells, with IC50s of 115.67 and 118.80 μM, respectively. It shows no cytotoxic effect at concentrations from 6.25-200 μM.
  • Phenolic acid
  • Exhibits NO inhibitory effects in vitro
  • Not cytotoxic at specified concentrations