Learn More
Medchemexpress LLC Methyl 5-hydroxypyridine-2-carboxylate | 30766-12-2 | 153.14 | 50 G
SDP
Catalog No. 5000449334
Encompass_Preferred
Methyl 5-hydroxypyridine-2-carboxylate is a phenolic acid found in the stems of Mahonia fortune. It demonstrates NO inhibitory effects in vitro, specifically inhibiting NO in LPS-stimulated RAW264.7 and BV2 cells with IC50s of 115.67 μM and 118.80 μM, respectively. The compound does not exhibit any cytotoxic effect at concentrations ranging from 6.25-200 μM.
  • Phenolic acid derived from Mahonia fortune
  • Exhibits NO inhibitory effects in vitro
  • No cytotoxic effects observed at tested concentrations
  • Suitable for research use only
Catalog No. 50-004-49334 Supplier Medchemexpress LLC Supplier No. HYW00596350G
Methyl 5-hydroxypyridine-2-carboxylate is a phenolic acid found in the stems of Mahonia fortune. It demonstrates NO inhibitory effects in vitro, specifically inhibiting NO in LPS-stimulated RAW264.7 and BV2 cells with IC50s of 115.67 μM and 118.80 μM, respectively. The compound does not exhibit any cytotoxic effect at concentrations ranging from 6.25-200 μM.
  • Phenolic acid derived from Mahonia fortune
  • Exhibits NO inhibitory effects in vitro
  • No cytotoxic effects observed at tested concentrations
  • Suitable for research use only