Learn More
Sigma Aldrich methyl-(5-methyl-thiazol-2-ylmethyl)-amine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2-methyl-1-hexene is a less reactive olefin that is hydroformylated in the presence of phosphite-modified rhodium catalysts
- The epoxidation of 2-methyl-1-hexene is catalyzed by human and rat p450 enzymes
Specifications
Specifications
| Molecular Formula | C6H10N2S |
| Linear Formula | C6H10N2S |
| MDL Number | MFCD08060729 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 142.22 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.