Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-N,4-O-Carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosylonate 98.0+%, TCI America™
SDP

Catalog No. M23291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23291G Supplier TCI America Supplier No. M23291G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 934591-79-4
Molecular Formula C17H21NO8S
Molecular Weight (g/mol) 399.414
InChI Key PWTWMNLOCAQKPJ-LDMYBJDZSA-N
Synonym Methyl (Phenyl 5-N,4-O-Carbonyl-3,5-dideoxy-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosid)onate
PubChem CID 133554242
IUPAC Name methyl 2-oxo-6-phenylsulfanyl-4-[(1S,2S)-1,2,3-trihydroxypropyl]-3a,4,7,7a-tetrahydro-3H-pyrano[3,4-d][1,3]oxazole-6-carboxylate
SMILES COC(=O)C1(CC2C(C(O1)C(C(CO)O)O)NC(=O)O2)SC3=CC=CC=C3

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 399.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-N,4-O-Carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosylonat
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.