Learn More
Polysciences, Inc. Nadic Methyl Anhydride (NMA)
SDP
Supplier:  Polysciences, Inc. 00886500
Encompass_Preferred
Liquid anhydride for curing epoxy resins. This material is a mixture of methyl isomers of methylbicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride. The larger the proportion of NMA used in the epoxy resin formula, the harder the resultant block.
Catalog No. NC1929760