Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl-5-norbornene-2,3-dicarboxylic anhydride

Catalog No. 2354311KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-543-11KG 1 kg
23-543-1250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-543-11KG Supplier Sigma Aldrich Supplier No. 2354311KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl-5-norbornene-2,3-dicarboxylic anhydride has been used:
  • As curing agent in the preparation of vertically aligned and penetrated carbon nanotube/polymer film
  • In the synthesis of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide

Specifications

CAS 25134-21-8
Density 1.232 g/mL (at 25°C (literature))
Molecular Formula C10H10O3
Refractive Index n20/D 1.506 (literature)
Linear Formula C10H10O3
Quantity 1 kg
Synonym Methyl nadic anhydride
Molecular Weight (g/mol) 178.18
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.