Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-Acetoxyhexanoate 98.0+%, TCI America™
SDP

Catalog No. A13495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13495G Supplier TCI America Supplier No. A13495G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104954-58-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00082930
InChI Key MFIRVMDIUPRJDB-UHFFFAOYSA-N
Synonym 6-Acetoxyhexanoic Acid Methyl Ester
PubChem CID 9979616
IUPAC Name methyl 6-(acetyloxy)hexanoate
SMILES COC(=O)CCCCCOC(C)=O

Specifications

Color Colorless
Quantity 5 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 6-Acetoxyhexanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.