Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-bromoindole-2-carboxylate, 97%

Catalog No. AAH6318706
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6318706 5 g
AAH6318703 1 g
AAH63187MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6318706 Supplier Thermo Scientific Chemicals Supplier No. H6318706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 372089-59-3 | C11H10BrNO2 | 268.11 g/mol

Methyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in acetone.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372089-59-3
MDL Number MFCD03094942
Synonym ethyl 6-bromoindole-2-carboxylate, 6-bromoindole-2-carboxylic acid ethyl ester, 6-bromo-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromo-2-indolecarboxylic ethyl ester, 6-bromo-2-ethoxylcarbonyl-1h-indole, 6-bromo-2-carbethoxyindole, 6-bromo-2-carboethoxyindole, 1h-indole-2-carboxylicacid, 6-bromo-, ethyl ester
PubChem CID 7009496
IUPAC Name ethyl 6-bromo-1H-indole-2-carboxylate

Specifications

Color White
Molecular Formula C10H8BrNO2
Quantity 5 g
Solubility Information Soluble in acetone.
InChI Key BUMYRKNZGPXFEG-UHFFFAOYSA-N
SMILES COC(=O)C1=CC2=CC=C(Br)C=C2N1
Molecular Weight (g/mol) 254.08
Formula Weight 254.08
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-bromoindole-2-carboxylate
Show More Show Less
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.