Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-Bromonicotinate 98.0+%, TCI America™
SDP

Catalog No. M26825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26825G 5 g
M26821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26825G Supplier TCI America Supplier No. M26825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26218-78-0
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD04972371
InChI Key NFLROFLPSNZIAH-UHFFFAOYSA-N
Synonym 6-Bromonicotinic Acid Methyl Ester, Methyl 6-Bromopyridine-3-carboxylate, 6-Bromopyridine-3-carboxylic Acid Methyl Ester
PubChem CID 13349623
IUPAC Name methyl 6-bromopyridine-3-carboxylate
SMILES COC(=O)C1=CC=C(Br)N=C1

Specifications

Melting Point 108°C
Color White
Quantity 5 g
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Bromonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.