Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-Fluoro-2-naphthoate 98.0+%, TCI America™
SDP

Catalog No. F04735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04735G 5 g
F04731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04735G Supplier TCI America Supplier No. F04735G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5043-00-5
Molecular Formula C12H9FO2
Molecular Weight (g/mol) 204.2
MDL Number MFCD06797106
InChI Key FXLVPMMENJMZBM-UHFFFAOYSA-N
Synonym 6-Fluoro-2-naphthoic Acid Methyl Ester
PubChem CID 44630303
IUPAC Name methyl 6-fluoronaphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)F

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 5 g
Formula Weight 204.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Fluoro-2-naphthoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.