Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-Fluoronicotinate 98.0+%, TCI America™
SDP

Catalog No. M27241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27241G 1 g
M27245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27241G Supplier TCI America Supplier No. M27241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1427-06-1
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD03095098
InChI Key HLYBWNNPVXFCPZ-UHFFFAOYSA-N
Synonym methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate
PubChem CID 13758765
IUPAC Name methyl 6-fluoropyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)F

Specifications

Melting Point 51°C
Color White-Yellow
Quantity 1 g
Formula Weight 155.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Fluoronicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.