Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-methylnicotinate, 97%

Catalog No. AC248210100
Change view
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC248210100 10 g Glass bottle
AC248210020 2 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC248210100 Supplier Thermo Scientific Chemicals Supplier No. 248210100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5470-70-2 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-70-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00006340
InChI Key VYPPZXZHYDSBSJ-UHFFFAOYSA-N
Synonym methyl 6-methylnicotinate, methyl-6-methylnicotinate, 6-methylnicotinic acid methyl ester, etoricoxib impurity e, unii-ya0k0cm5pk, methyl 6-methyl nicotinate, 6-methyl-nicotinic acid methyl ester, 6-methyl nicotinic acid methyl ester, ya0k0cm5pk, 3-methoxycarbonyl-6-methylpyridine
PubChem CID 231548
IUPAC Name methyl 6-methylpyridine-3-carboxylate
SMILES CC1=NC=C(C=C1)C(=O)OC

Specifications

Melting Point 31.0°C to 37.0°C
Color Colorless to Yellow
Boiling Point 108.0°C to 110.0°C (22.5 mmHg)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 151.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 6-methylnicotinate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.