Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 7-chloroheptanoate, 98%, Thermo Scientific™

Catalog No. AAH2709814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2709814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2709814 Supplier Thermo Scientific Chemicals Supplier No. H2709814

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26040-62-0
Molecular Formula C8H15ClO2
Molecular Weight (g/mol) 178.66
MDL Number MFCD09751145
InChI Key YKSFGXAJDCLJKE-UHFFFAOYSA-N
Synonym 7-chloroheptanoic acid, methyl ester, heptanoic acid, 7-chloro-, methyl ester, acmc-20akfu, methyl 7-chloroheptanoate, 7-chloroheptanoic acid methyl ester, heptanoic acid, 7-chloro, methyl ester
PubChem CID 529989
IUPAC Name methyl 7-chloroheptanoate
SMILES COC(=O)CCCCCCCl

Specifications

Refractive Index 1.4425
Linear Formula Cl(CH2)6CO2CH3
Quantity 25 g
Formula Weight 178.66
Percent Purity 98%
Chemical Name or Material Methyl 7-chloroheptanoate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.