Learn More
Medchemexpress LLC Methyl 8-(tert-butyl)-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate | 1690176-75-0 | MFCD28144723 | 99.8% | 267.71 g/mol | C12H14ClN3O2 | 5 MG
SDP
Catalog No. 5000222646
Encompass_Preferred
PAR-2-IN-1 is a small-molecule inhibitor of the protease-activated receptor 2 (PAR2) signaling pathway with reported anti-inflammatory and anticancer activity. It is supplied for research use and is available as a solid powder and as a 10 mM solution in DMSO. The compound has high reported purity and a confirmed molecular formula suitable for biological and analytical assays.
Catalog No. 50-002-22646 Supplier Medchemexpress LLC Supplier No. HY1385585MG
PAR-2-IN-1 is a small-molecule inhibitor of the protease-activated receptor 2 (PAR2) signaling pathway with reported anti-inflammatory and anticancer activity. It is supplied for research use and is available as a solid powder and as a 10 mM solution in DMSO. The compound has high reported purity and a confirmed molecular formula suitable for biological and analytical assays.