Learn More
Medchemexpress LLC Methyl 8-(tert-butyl)-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate | 1690176-75-0 | MFCD28144723 | 99.8% | C12H14ClN3O2 | 10MG
SDP
Catalog No. 5000260296
Encompass_Preferred
PAR-2-IN-1 is a small-molecule inhibitor of protease-activated receptor 2 (PAR2) signaling with reported anti-inflammatory and anticancer effects. The compound is supplied as a solid research reagent with defined solubility in DMSO and formulation examples for in vivo studies.
  • Selective PAR2 pathway inhibitor.
  • Demonstrated anti-inflammatory and anticancer activity in preclinical studies.
  • High purity (99.8%).
  • Soluble in DMSO (~83.33 mg/mL) and formulatable for in vivo use.
  • Supplied as a stable powder with recommended storage at -20°C.
  • Available in multiple research quantities.
Catalog No. 50-002-60296 Supplier Medchemexpress LLC Supplier No. HY13855810MG
PAR-2-IN-1 is a small-molecule inhibitor of protease-activated receptor 2 (PAR2) signaling with reported anti-inflammatory and anticancer effects. The compound is supplied as a solid research reagent with defined solubility in DMSO and formulation examples for in vivo studies.
  • Selective PAR2 pathway inhibitor.
  • Demonstrated anti-inflammatory and anticancer activity in preclinical studies.
  • High purity (99.8%).
  • Soluble in DMSO (~83.33 mg/mL) and formulatable for in vivo use.
  • Supplied as a stable powder with recommended storage at -20°C.
  • Available in multiple research quantities.