Learn More
Medchemexpress LLC Methyl 9(Z),11(E)-octadecadienoate | 13058-52-1 | 99.2% | 294.47 g/mol | C19H34O2 | 5MG
SDP
Catalog No. 5000239662
Encompass_Preferred
Methyl 9(Z),11(E)-octadecadienoate is the methyl ester of conjugated linoleic acid, supplied as a high-purity analytical reagent for lipid and fatty-acid research and method development. It is commonly used as a reference standard for chromatographic calibration, isomer identification, and mechanistic studies of fatty acids. The material is provided in small milligram quantities with supporting documentation for safe handling and analytical use.
  • High purity 99.2%.
  • Suitable as an analytical reference standard for lipid and fatty-acid research.
  • Provided in milligram pack sizes for small-scale use.
  • Includes a product data sheet and a safety data sheet.
  • Chemical identifiers: CAS 13058-52-1; formula C19H34O2; Mw 294.5 g/mol.
Catalog No. 50-002-39662 Supplier Medchemexpress LLC Supplier No. HY1243705MG
Methyl 9(Z),11(E)-octadecadienoate is the methyl ester of conjugated linoleic acid, supplied as a high-purity analytical reagent for lipid and fatty-acid research and method development. It is commonly used as a reference standard for chromatographic calibration, isomer identification, and mechanistic studies of fatty acids. The material is provided in small milligram quantities with supporting documentation for safe handling and analytical use.
  • High purity 99.2%.
  • Suitable as an analytical reference standard for lipid and fatty-acid research.
  • Provided in milligram pack sizes for small-scale use.
  • Includes a product data sheet and a safety data sheet.
  • Chemical identifiers: CAS 13058-52-1; formula C19H34O2; Mw 294.5 g/mol.