Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 9H-xanthene-9-carboxylate | 39497-06-8 | 95.0% | C15H12O3 | 250 MG
SDP

Catalog No. 5000358379
Encompass_Preferred

Methyl 9H-xanthene-9-carboxylate (Methyl xanthanoate) is a drug intermediate used for the synthesis of various active compounds.

  • Drug intermediate
  • Used for the synthesis of various active compounds
  • Appearance: light yellow to yellow oil
  • Stable under recommended storage conditions
  • Long-term storage for pure form: -20°C for 3 years; 4°C for 2 years
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-58379 Supplier Medchemexpress LLC Supplier No. 250MGHY78996
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 9H-xanthene-9-carboxylate (Methyl xanthanoate) is a drug intermediate used for the synthesis of various active compounds.

  • Drug intermediate
  • Used for the synthesis of various active compounds
  • Appearance: light yellow to yellow oil
  • Stable under recommended storage conditions
  • Long-term storage for pure form: -20°C for 3 years; 4°C for 2 years
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.