Learn More
Selleck Chemical LLC Methyl a-D-mannopyranoside S5047-25mg
SDP
Catalog No. 503108505
Encompass_Preferred
Methyl -D-mannopyranoside can be used as an intermediate for chemical sythesis and is a competitor inhibitor of the binding of mannose by Escherichia coli
Catalog No. 50-310-8505 Supplier Selleck Chemical LLC Supplier No. S504725MG
Methyl -D-mannopyranoside can be used as an intermediate for chemical sythesis and is a competitor inhibitor of the binding of mannose by Escherichia coli