Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Acetate (Certified ACS), Fisher Chemical™

Catalog No. M2034 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M203-4 4 L Glass Bottle
M203-500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. M203-4
This item is not returnable. View return policy

Chemical Identifiers

CAS 79-20-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00008711
InChI Key KXKVLQRXCPHEJC-UHFFFAOYSA-N
Synonym tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
PubChem CID 6584
ChEBI CHEBI:77700
IUPAC Name methyl acetate
SMILES CC(=O)OC

Specifications

Boiling Range 1.0°C max.
Melting Point -98°C
Color Colorless
Density 0.930g/cm³
Boiling Point 57.4°C
Flash Point −26°C
Packaging Glass Bottle
Linear Formula CH3COOCH3
Quantity 4 L
UN Number 1231
Residue after Evaporation 0.002% max.
Specific Gravity 0.933 to 0.938 (at 20°C)
Formula Weight 74.08
Grade Certified
Physical Form Liquid
Chemical Name or Material Methyl Acetate
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.