Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl acetate ReagentPlus
Catalog No. 502898865
Encompass
Methyl acetate is an aliphatic ester that can be prepared via carbonylation of dimethyl ether over zeolites. MA is formed as a by-product during the preparation of polyvinyl alcohol from acetic acid and methanol.
Catalog No. 50-289-8865 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1863254L
Methyl acetate is an aliphatic ester that can be prepared via carbonylation of dimethyl ether over zeolites. MA is formed as a by-product during the preparation of polyvinyl alcohol from acetic acid and methanol.