Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl acetate ReagentPlus
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 18632518LCS
Encompass
Methyl acetate is an aliphatic ester that can be prepared via carbonylation of dimethyl ether over zeolites. MA is formed as a by-product during the preparation of polyvinyl alcohol from acetic acid and methanol.
Catalog No. 50-289-8896