Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl acetate ReagentPlus1L
Catalog No. 502897671
Encompass
Methyl acetate is an aliphatic ester that can be prepared via carbonylation of dimethyl ether over zeolites. MA is formed as a by-product during the preparation of polyvinyl alcohol from acetic acid and methanol.
Catalog No. 50-289-7671 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1863251L
Methyl acetate is an aliphatic ester that can be prepared via carbonylation of dimethyl ether over zeolites. MA is formed as a by-product during the preparation of polyvinyl alcohol from acetic acid and methanol.