Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl acetoacetate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019794 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9794 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9794 Supplier MilliporeSigma Supelco Supplier No. 146105MLF

missing translation for 'validMainframeMsg'

Specifications

CAS 105-45-3
Ignition Point 536°F
Density 1.076 g/mL (at 25°C)
Flash Point 70°C
Physical Form Neat
Linear Formula CH3 COCH2 COOCH3
MDL Number MFCD00008784
Melting Point -80°C
Boiling Point 169°C to 170°C (70 mmHg)
Synonym 3-Oxobutanoic acid methyl ester; Acetoacetic acid methyl ester
Type Analytical Standard
Quantity 5 mL
Formula Weight 116.12
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.