Learn More
Medchemexpress LLC Methyl acetylacetate | 105-45-3 | MFCD00008784 | 99.9% | 116.12 g/mol | C5H8O3 | 250g
SDP
Catalog No. 5000266651
Encompass_Preferred
Methyl acetylacetate is a chemical reagent used in the synthesis of pharmaceuticals for the synthesis of -substituted acetoacetate and cyclic compounds such as pyrazole pyrimidine and coumarin derivatives Methyl acetoacetate can be used in the study of inflammatory diseases[1][2][3]
Catalog No. 50-002-66651 Supplier Medchemexpress LLC Supplier No. HYY1298250G
Methyl acetylacetate is a chemical reagent used in the synthesis of pharmaceuticals for the synthesis of -substituted acetoacetate and cyclic compounds such as pyrazole pyrimidine and coumarin derivatives Methyl acetoacetate can be used in the study of inflammatory diseases[1][2][3]