Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl acrylate

Catalog No. 503124390 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
2 L
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-4390 2 L
50-312-4388 1 L
50-312-4391 18 L
50-312-4389 250 mL
50-312-4392 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-312-4390 Supplier Sigma Aldrich Supplier No. M273012L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl acrylate

Specifications

CAS 96-33-3
Melting Point -75°C
Density 0.956 g/mL (at 25°C)
Boiling Point 80°C
Molecular Formula C4H6O2
Refractive Index n20/D 1.402 (literature)
Linear Formula CH2=CHCOOCH3
Quantity 2 L
MDL Number MFCD00008627
Molecular Weight (g/mol) 86.09
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.