Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl acrylate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019988 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9988 1 mL
11-101-8698 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9988 Supplier MilliporeSigma Supelco Supplier No. 767781MLF
Add to Cart
Add to Cart
Environmental and Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 874°F
CAS 96-33-3
Vapor Pressure 67.5 mmHg (20°C)
Vapor Density 3 (vs. air)
Density 0.956 g/mL (at 25°C)
Flash Point -3°C - Closed Cup
Physical Form Neat
Linear Formula CH2=CHCOOCH3
MDL Number MFCD00008627
Melting Point -75°C
Boiling Point 80°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 86.09
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.