Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl all-cis-5,8,11,14,17-eicosapentaenoate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013793 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3793 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3793 Supplier MilliporeSigma Supelco Supplier No. 17266100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl all-cis-5,8,11,14,17-eicosapentaenoate is a fatty acid methyl ester.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 2734-47-6
Flash Point 104°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2 CH=CH)5(CH2)3 CO2 CH3
MDL Number MFCD02259226
Synonym cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 316.48
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.