Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl alpha-bromoacrylate

Catalog No. 5884661G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-846-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58-846-61G Supplier Sigma Aldrich Supplier No. 5884661G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl α-bromoacrylate can undergo radical copolymerization with vinyl triacetoxysilane (VTAS) in the presence of azobisisobutyronitrile as the initiator. Dibromocyclopropanation of methyl α-bromoacrylate can yield methyl 1,1,2-tribromocyclopropanecarboxylate. It may be reduced with zinc dust and deuterium oxide to synthesize. Methyl acrylate-α-d with a high isotopic purity. These may undergo conjugate addition with methoxide, thiolates, oxime anions, hydrazones and EtZnCl to afford ester enolates.

Specifications

CAS 4519-46-4
Density 1.556 g/mL (at 25°C)
Boiling Point 70°C to 75°C (48 mmHg)
Molecular Formula C4H5BrO2
Linear Formula C4H5BrO2
Quantity 1 g
MDL Number MFCD08276754
Molecular Weight (g/mol) 164.99
Percent Purity 95%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.