Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl alpha-D-galactopyranoside monohydrate, Thermo Scientific™

Catalog No. AAL0804906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0804906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0804906 Supplier Thermo Scientific Chemicals Supplier No. L0804906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl alpha-D-galactopyranoside monohydrate is used in medicine, industry.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34004-14-3
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00150748
InChI Key HOVAGTYPODGVJG-PZRMXXKTSA-N
Synonym methyl alpha-d-galactopyranoside monohydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol hydrate, methyl-alpha-d-galactopyranoside, methyl ?-galactoside hydrate, methyl a-d-galactopyranoside,monohydrate, methyl alpha-d-galactopyranoside hydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxyoxane-3,4,5-triol hydrate
PubChem CID 2724703
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol;hydrate
SMILES CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 105°C to 108°C
Quantity 5 g
Beilstein 81570
Solubility Information Insoluble in water.
Optical Rotation +169° (c=1.5 in Methanol)
Formula Weight 212.20 (194.18 Anhydrous)
Chemical Name or Material Methyl alpha-D-galactopyranoside monohydrate
EINECSNumber 222-251-7
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.