Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl alpha-D-mannopyranoside | 617-04-9 | MFCD00063262 | >98.0% | 194.18 g·mol⁻¹ | C7H14O6 | 100 G
SDP

Catalog No. 5000299450
Encompass_Preferred

Methyl α-D-mannopyranoside is a methylated monosaccharide research reagent used to study carbohydrate-protein interactions and to inhibit mannose-specific lectin binding. It has been examined as a macrophage-targeting agent for anti-tuberculosis inhalation therapy and is supplied as a high-purity solid for biochemical and analytical applications.

  • High purity (>98.0% by HPLC).
  • Molecular formula C7H14O6; molecular weight 194.18 g·mol⁻¹.
  • CAS 617-04-9.
  • White to off-white powder or crystals.
  • Useful as a lectin inhibitor and carbohydrate-protein interaction probe.
  • Investigated for macrophage-targeting in inhalation therapy.
  • Available in laboratory-scale pack sizes suitable for research.

Catalog No. 50-002-99450 Supplier Medchemexpress LLC Supplier No. HYW039897100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl α-D-mannopyranoside is a methylated monosaccharide research reagent used to study carbohydrate-protein interactions and to inhibit mannose-specific lectin binding. It has been examined as a macrophage-targeting agent for anti-tuberculosis inhalation therapy and is supplied as a high-purity solid for biochemical and analytical applications.

  • High purity (>98.0% by HPLC).
  • Molecular formula C7H14O6; molecular weight 194.18 g·mol⁻¹.
  • CAS 617-04-9.
  • White to off-white powder or crystals.
  • Useful as a lectin inhibitor and carbohydrate-protein interaction probe.
  • Investigated for macrophage-targeting in inhalation therapy.
  • Available in laboratory-scale pack sizes suitable for research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.