Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl alpha-L-Fucopyranoside 98.0+%, TCI America™
SDP

Catalog No. M10511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10511G Supplier TCI America Supplier No. M10511G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monosaccharide used as a drug intermediate in research applications.

Chemical Identifiers

CAS 14687-15-1
Molecular Formula C7H14O5
Molecular Weight (g/mol) 178.184
MDL Number MFCD00069802
InChI Key OHWCAVRRXKJCRB-CXNFULCWSA-N
PubChem CID 446578
IUPAC Name (2R,3S,4R,5S,6S)-2-methoxy-6-methyloxane-3,4,5-triol
SMILES CC1C(C(C(C(O1)OC)O)O)O

Specifications

Melting Point 159°C
Color White
Quantity 1 g
Formula Weight 178.18
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Methyl alpha-L-Fucopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.