Learn More
Selleck Chemical LLC Methyl Aminolevulinate Hydrochloride S5372-25mg
SDP
Catalog No. 503111521
Encompass_Preferred
Methyl Aminolevulinate (Aminolevulinic acid methyl ester Hydrochloride Methyl 5-aminolevulinate Hydrochloride) is a prodrug that is metabolised to protoporphyrin IX used in photodynamic therapy
Catalog No. 50-311-1521 Supplier Selleck Chemical LLC Supplier No. S537225MG
Methyl Aminolevulinate (Aminolevulinic acid methyl ester Hydrochloride Methyl 5-aminolevulinate Hydrochloride) is a prodrug that is metabolised to protoporphyrin IX used in photodynamic therapy