Learn More
Medchemexpress LLC Methyl aminolevulinate hydrochloride | 79416-27-6 | 99.0% | C6H12ClNO3 | 10 G
SDP
Catalog No. 5000432699
Encompass_Preferred
Methyl aminolevulinate hydrochloride is an agent used as a sensitizer in photodynamic therapy (PDT). It acts as a proagent that can be metabolized to Protoporphyrin IX. This product is for research use only.
  • Utilized as a sensitizer in photodynamic therapy (PDT)
  • Acts as a proagent
  • Metabolized to Protoporphyrin IX
Catalog No. 50-004-32699 Supplier Medchemexpress LLC Supplier No. HYA0169A10G
Add to Cart
Add to Cart
Methyl aminolevulinate hydrochloride is an agent used as a sensitizer in photodynamic therapy (PDT). It acts as a proagent that can be metabolized to Protoporphyrin IX. This product is for research use only.
  • Utilized as a sensitizer in photodynamic therapy (PDT)
  • Acts as a proagent
  • Metabolized to Protoporphyrin IX