Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl aminolevulinate hydrochloride | 79416-27-6 | 99.0% | C6H12ClNO3 | 5 G
SDP

Catalog No. 5000432244
Encompass_Preferred

Methyl aminolevulinate hydrochloride is an agent used as a sensitizer in photodynamic therapy (PDT). Methyl aminolevulinate is a proagent that can be metabolized to Protoporphyrin IX.

  • Used as a sensitizer in photodynamic therapy (PDT)
  • Metabolized to Protoporphyrin IX
  • Molecular weight: 181.62
  • Appearance: White to off-white solid
  • Soluble in H2O at 100 mg/mL
  • Soluble in DMSO at 100 mg/mL
  • For research use only

Catalog No. 50-004-32244 Supplier Medchemexpress LLC Supplier No. HYA0169A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl aminolevulinate hydrochloride is an agent used as a sensitizer in photodynamic therapy (PDT). Methyl aminolevulinate is a proagent that can be metabolized to Protoporphyrin IX.

  • Used as a sensitizer in photodynamic therapy (PDT)
  • Metabolized to Protoporphyrin IX
  • Molecular weight: 181.62
  • Appearance: White to off-white solid
  • Soluble in H2O at 100 mg/mL
  • Soluble in DMSO at 100 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.