Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl aminomethanimidothioate hydroiodide, 95%, Thermo Scientific™

Catalog No. KM00619EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM00619EA 10 g Amber Glass Bottle
KM00619DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. KM00619EA Supplier Thermo Scientific Chemicals Supplier No. KM00619EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4338-95-8
Molecular Formula C2H7IN2S
Molecular Weight (g/mol) 218.06
MDL Number MFCD00035598
InChI Key LFXAECSQJSRSTP-UHFFFAOYSA-N
Synonym methyl aminomethanimidothioate hydroiodide, sulfiode, methylthiouronium iodide, methyl carbamimidothioate hydroiodide, s-methylthiuronium iodide, s-methylthiouronium iodide, s-methylisothiouronium iodide, 2-methyl-2-thiopseudourea hydroiodide, 2-methylisothiouronium iodide, s-methylisothiourea hydriodide
PubChem CID 197812
SMILES [H+].[I-].CSC(N)=N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name methyl carbamimidothioate;hydroiodide
Formula Weight 218.06
Percent Purity 95%
Chemical Name or Material Methyl aminomethanimidothioate hydroiodide
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.