Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl anisate | 121-98-2 | MFCD00008437 | 99.3% | 166.18 g·mol⁻¹ | C9H10O3 | 1 ML
SDP

Catalog No. 5000303262
Encompass_Preferred

Methyl anisate is an endogenous metabolite (methyl 4-methoxybenzoate) used as an analytical reagent and reference standard. It is available as a 10 mM solution in DMSO (1 mL) or as solid material, with reported purity near 99% and a molecular weight of 166.18 g·mol⁻¹. Typical applications include analytical characterization and laboratory research assays.

  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High purity suitable for analytical and research use.
  • Molecular weight 166.18 g·mol⁻¹, chemical formula C9H10O3.
  • Appearance: white to light yellow solid; solution supplied in DMSO.
  • Storage recommendations provided for powder and solutions to maintain stability.

Catalog No. 50-003-03262 Supplier Medchemexpress LLC Supplier No. HYW01534210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl anisate is an endogenous metabolite (methyl 4-methoxybenzoate) used as an analytical reagent and reference standard. It is available as a 10 mM solution in DMSO (1 mL) or as solid material, with reported purity near 99% and a molecular weight of 166.18 g·mol⁻¹. Typical applications include analytical characterization and laboratory research assays.

  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High purity suitable for analytical and research use.
  • Molecular weight 166.18 g·mol⁻¹, chemical formula C9H10O3.
  • Appearance: white to light yellow solid; solution supplied in DMSO.
  • Storage recommendations provided for powder and solutions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.