Learn More
Medchemexpress LLC Methyl arachidate (methyl icosanoate) | 1120-28-1 | MFCD00009014 | 326.56 | 5 G
SDP
Catalog No. 5000445891
Encompass_Preferred
Methyl arachidate (Methyl icosanoate) is a component of lipid mixtures. It can be isolated from flowering plants, inhibits LTA4H, and shows no antifeedant activity against *Leptinotarsa decemlineata*.
  • Component of lipid mixtures
  • Can be isolated from flowering plants
  • Inhibits LTA4H
  • Shows no antifeedant activity against *Leptinotarsa decemlineata*
  • Solid, white to off-white appearance
  • Molecular formula C21H42O2
Catalog No. 50-004-45891 Supplier Medchemexpress LLC Supplier No. HYW0042915G
Methyl arachidate (Methyl icosanoate) is a component of lipid mixtures. It can be isolated from flowering plants, inhibits LTA4H, and shows no antifeedant activity against *Leptinotarsa decemlineata*.
  • Component of lipid mixtures
  • Can be isolated from flowering plants
  • Inhibits LTA4H
  • Shows no antifeedant activity against *Leptinotarsa decemlineata*
  • Solid, white to off-white appearance
  • Molecular formula C21H42O2