Learn More
Medchemexpress LLC HY-101461 500mg , Methyl-β-cyclodextrin CAS:128446-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101461500MG
Encompass_Preferred
HY-101461 500mg Methyl-β-cyclodextrin CAS: 128446-36-6 Methyl-β-cyclodextrin (Methyl-beta-cyclodextrin) is a cyclic heptasaccharide used to deliver hydrophobic drugs based on its property of solubilizing non-polar substances. Methyl-β-cyclodextrin is also extensively used as a cholesterol-depleting reagent. Methyl-β-cyclodextrin strongly reduces clathrin-dependent endocytosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9596