Learn More
Selleck Chemical LLC Methyl-b-cyclodextrin
SDP
Catalog No. 5000002527
Encompass_Preferred
Methyl- -cyclodextrin (M CD) is a cholesterol-removing agent mainly used for lipid raft disruption Methyl- -cyclodextrin induces caspase-dependent apoptosis in PEL cells and inhibits the growth and invasion of PEL cells without apparent adverse effects suggesting the potential effective antitumor activities
Catalog No. 50-000-02527 Supplier Selleck Chemical LLC Supplier No. S6827500MG
Add to Cart
Add to Cart
Methyl- -cyclodextrin (M CD) is a cholesterol-removing agent mainly used for lipid raft disruption Methyl- -cyclodextrin induces caspase-dependent apoptosis in PEL cells and inhibits the growth and invasion of PEL cells without apparent adverse effects suggesting the potential effective antitumor activities