Learn More
Selleck Chemical LLC Methyl-b-cyclodextrin
SDP
Supplier:  Selleck Chemical LLC S68271G
Encompass_Preferred
Methyl- -cyclodextrin (M CD) is a cholesterol-removing agent mainly used for lipid raft disruption Methyl- -cyclodextrin induces caspase-dependent apoptosis in PEL cells and inhibits the growth and invasion of PEL cells without apparent adverse effects suggesting the potential effective antitumor activities
Catalog No. 50-000-02594