Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl B-D-glucopyra 10g | 709-50-2 | 194.18 | C7H14O6 | 10 G
SDP

Catalog No. 5000308263
Encompass_Preferred

Methyl β-D-glucopyranoside is a methylated glucose derivative supplied as a white solid research reagent. It is used as a glycosyl donor and analytical standard in carbohydrate chemistry, enzymatic glycosylation, and biocatalysis studies.

  • High purity (99.91%) suitable for research applications.
  • Molecular weight 194.18 g/mol.
  • Chemical formula C7H14O6.
  • White solid, easy to weigh and dissolve in common solvents.
  • Stable when stored sealed at 4°C; follow the SDS for handling and storage.

Catalog No. 50-003-08263 Supplier Medchemexpress LLC Supplier No. HY11628410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl β-D-glucopyranoside is a methylated glucose derivative supplied as a white solid research reagent. It is used as a glycosyl donor and analytical standard in carbohydrate chemistry, enzymatic glycosylation, and biocatalysis studies.

  • High purity (99.91%) suitable for research applications.
  • Molecular weight 194.18 g/mol.
  • Chemical formula C7H14O6.
  • White solid, easy to weigh and dissolve in common solvents.
  • Stable when stored sealed at 4°C; follow the SDS for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.