Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl B-D-glucopyra 10mM 1mL | 709-50-2 | 194.18 g/mol | C7H14O6 | 1 ML
SDP

Catalog No. 5000227974
Encompass_Preferred

Methyl β-D-glucopyranoside is a methyl glycoside reagent commonly used in organic synthesis and biocatalysis research. It is supplied as a ready-to-use 10 mM aqueous solution (1 mL) and is also available in multiple solid sizes for preparative use.

  • Purity 99.91%.
  • Molecular weight 194.18 g/mol.
  • Chemical formula C7H14O6.
  • CAS number 709-50-2.
  • Storage: 4°C, sealed and away from moisture; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-27974 Supplier Medchemexpress LLC Supplier No. HY11628410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Methyl β-D-glucopyranoside is a methyl glycoside reagent commonly used in organic synthesis and biocatalysis research. It is supplied as a ready-to-use 10 mM aqueous solution (1 mL) and is also available in multiple solid sizes for preparative use.

  • Purity 99.91%.
  • Molecular weight 194.18 g/mol.
  • Chemical formula C7H14O6.
  • CAS number 709-50-2.
  • Storage: 4°C, sealed and away from moisture; in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.