Learn More
Medchemexpress LLC Methyl B-D-glucopyra 1g | 709-50-2 | 194.18 g/mol | C7H14O6 | 1 G
SDP
Catalog No. 5000227337
Encompass_Preferred
Methyl β-D-glucopyranoside is a methyl glycoside derivative of D-glucose used as a model glycosyl acceptor in enzymatic glycosylation studies and as an analytical standard for carbohydrate research. It is typically supplied as a white crystalline solid with high reported purity, and is soluble in water for solution-based assays.
  • High purity suitable for research and analytical use.
  • Commonly used as a model glycosyl acceptor in enzymatic glycosylation reactions.
  • White crystalline solid form for accurate weighing and storage.
  • Molecular weight 194.18 g/mol and formula C7H14O6 for precise stoichiometry.
  • Available in small pack sizes suitable for laboratory experiments.
Catalog No. 50-002-27337 Supplier Medchemexpress LLC Supplier No. HY1162841G
Methyl β-D-glucopyranoside is a methyl glycoside derivative of D-glucose used as a model glycosyl acceptor in enzymatic glycosylation studies and as an analytical standard for carbohydrate research. It is typically supplied as a white crystalline solid with high reported purity, and is soluble in water for solution-based assays.
  • High purity suitable for research and analytical use.
  • Commonly used as a model glycosyl acceptor in enzymatic glycosylation reactions.
  • White crystalline solid form for accurate weighing and storage.
  • Molecular weight 194.18 g/mol and formula C7H14O6 for precise stoichiometry.
  • Available in small pack sizes suitable for laboratory experiments.