Learn More
Medchemexpress LLC Methyl B-D-glucopyra 25g | 709-50-2 | 194.18 g/mol | C7H14O6 | 25 G
SDP
Catalog No. 5000229855
Encompass_Preferred
Methyl β-D-glucopyranoside is the methyl glycoside derivative of D-glucose supplied as a white to off-white solid and used as an analytical reference standard and substrate in glycoscience and enzymatic synthesis research. The compound has molecular formula C7H14O6, molecular weight 194.18 g/mol, and is typically supplied at 99.91% purity.
  • High purity (99.91% by HPLC) suitable for analytical use.
  • White to off-white solid, easy to weigh and dissolve.
  • Stable when stored sealed at 4°C away from moisture.
  • Used as a substrate in glycoscience and enzymatic assays.
  • Available in common lab pack sizes, including 25 g.
Catalog No. 50-002-29855 Supplier Medchemexpress LLC Supplier No. HY11628425G
Methyl β-D-glucopyranoside is the methyl glycoside derivative of D-glucose supplied as a white to off-white solid and used as an analytical reference standard and substrate in glycoscience and enzymatic synthesis research. The compound has molecular formula C7H14O6, molecular weight 194.18 g/mol, and is typically supplied at 99.91% purity.
  • High purity (99.91% by HPLC) suitable for analytical use.
  • White to off-white solid, easy to weigh and dissolve.
  • Stable when stored sealed at 4°C away from moisture.
  • Used as a substrate in glycoscience and enzymatic assays.
  • Available in common lab pack sizes, including 25 g.