Learn More
Medchemexpress LLC Methyl behenate | 929-77-1 | 99.8% | 354.62 | 10 G
SDP
Catalog No. 5000453096
Encompass_Preferred
Methyl behenate (Methyl docosanoate) is a fatty acid methyl ester. It can be isolated from the plant Aspidopterys obcordata Lemsl and can be used as a phase change material.
  • Fatty acid methyl ester
  • Can be isolated from Aspidopterys obcordata Lemsl
  • Can be used as a phase change material
Catalog No. 50-004-53096 Supplier Medchemexpress LLC Supplier No. HYW00908210G
Methyl behenate (Methyl docosanoate) is a fatty acid methyl ester. It can be isolated from the plant Aspidopterys obcordata Lemsl and can be used as a phase change material.
  • Fatty acid methyl ester
  • Can be isolated from Aspidopterys obcordata Lemsl
  • Can be used as a phase change material