Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl behenate | 929-77-1 | 99.8% | 354.62 | 100 G
SDP

Catalog No. 5000453173
Encompass_Preferred

Methyl behenate (Methyl docosanoate) is a fatty acid methyl ester that can be isolated from the plant Aspidopterys obcordata Lemsl. It is commonly used as a phase change material.

  • Solid appearance
  • White to off-white color
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
  • Soluble in ethanol at 1.25 mg/mL (3.52 mM; needs ultrasonic)

Catalog No. 50-004-53173 Supplier Medchemexpress LLC Supplier No. HYW009082100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl behenate (Methyl docosanoate) is a fatty acid methyl ester that can be isolated from the plant Aspidopterys obcordata Lemsl. It is commonly used as a phase change material.

  • Solid appearance
  • White to off-white color
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
  • Soluble in ethanol at 1.25 mg/mL (3.52 mM; needs ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.