Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl behenate, 99%, analytical standard for GC

Catalog No. AC226810050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226810050 5 g
AC226810010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226810050 Supplier Thermo Scientific Chemicals Supplier No. 226810050
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 929-77-1 | C23H46O2 | 354.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-77-1
Molecular Formula C23H46O2
Molecular Weight (g/mol) 354.62
MDL Number MFCD00009347
InChI Key QSQLTHHMFHEFIY-UHFFFAOYSA-N
Synonym methyl behenate, docosanoic acid, methyl ester, behenic acid methyl ester, docosanoic acid methyl ester, behenic acid, methyl ester, unii-04kbo9r771, behenic methyl ester, kemester 9022, docosanoic acid methyl, acmc-209rjf
PubChem CID 13584
IUPAC Name methyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 54.0°C to 56.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)20CO2CH3
Quantity 5 g
Merck Index 15,1022
Solubility Information (5% in Hexane) Clear colorless solution
Formula Weight 354.61
Percent Purity 99%
Chemical Name or Material Methyl behenate
EINECSNumber 213-207-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.