Learn More
Medchemexpress LLC Methyl behenate (methyl docosanoate) | 929-77-1 | 99.8% | 354.62 | 500 MG
SDP
Catalog No. 5000451224
Encompass_Preferred
Methyl behenate (Methyl docosanoate) is a fatty acid methyl ester. Methyl behenate can be isolated from the plant Aspidopterys obcordata Hemsl. It can be used as a phase change material.
  • Fatty acid methyl ester.
  • Can be isolated from the plant Aspidopterys obcordata Hemsl.
  • Can be used as a phase change material.
Catalog No. 50-004-51224 Supplier Medchemexpress LLC Supplier No. HYW009082500MG
Methyl behenate (Methyl docosanoate) is a fatty acid methyl ester. Methyl behenate can be isolated from the plant Aspidopterys obcordata Hemsl. It can be used as a phase change material.
  • Fatty acid methyl ester.
  • Can be isolated from the plant Aspidopterys obcordata Hemsl.
  • Can be used as a phase change material.