Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Benzimidazole-5-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M25821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25821G Supplier TCI America Supplier No. M25821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26663-77-4
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD03407310
InChI Key WJHHIVYNOVTVGY-UHFFFAOYSA-N
Synonym methyl benzimidazole-5-carboxylate, methyl 1h-benzimidazole-5-carboxylate, methyl 1h-benzo d imidazole-5-carboxylate, methyl 1h-1,3-benzodiazole-5-carboxylate, 1h-benzimidazole-5-carboxylicacidmethylester, 1h-benzimidazole-5-carboxylic acid methyl ester, methyl 1h-benzo d imidazole-6-carboxylate, benzimidazole-5-carboxylic acid methyl ester, 1h-benzoimidazole-5-carboxylic acid methyl ester, 1h-benzimidazole-5-carboxylic acid, methyl ester
PubChem CID 2779733
IUPAC Name methyl 3H-benzimidazole-5-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)N=CN2

Specifications

Melting Point 141°C
Quantity 1 g
Formula Weight 176.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Benzimidazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.